1-(1-Naphthyl)-2-thiourea 97%

Danger

Synonyms: N-(1-Naphthyl)thiourea Antu

A19024-09 A19024-18 A19024-18 A19024-30 A19024-30 A19024-09
AAA19024-30EA 613.83 USD
AAA19024-30 CAAAA19024-09 CAAAA19024-18 AAA19024-09 CAAAA19024-30 AAA19024-18
1-(1-Naphthyl)-2-thiourea 97%
1-(1-Naphthyl)-2-thiourea
Formula: C₁₀H₇NHC(S)NH₂
MW: 202.28 g/mol
Boiling Pt: 360 °C (1013 hPa)
Melting Pt: 185…191 °C
Density: >1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00041824
CAS Number: 86-88-4
EINECS: 201-706-3
UN: 1651
ADR: "6.1",II
Merck Index: 13,00729

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR