2-Methyl-6-quinolinol 98%

Warning

Synonyms: 6-Hydroxy-2-methylquinoline

A18846-14 A18846-03 A18846-14 A18846-06 A18846-03 A18846-06
AAA18846-06EA 357.7 USD
AAA18846-06 CAAAA18846-14 CAAAA18846-03 CAAAA18846-06 AAA18846-14 AAA18846-03
2-Methyl-6-quinolinol 98%
2-Methyl-6-quinolinol
Formula: C₁₀H₉NO
MW: 159.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00047600
CAS Number: 613-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR