Saponin

Supplier: Thermo Scientific Chemicals
Warning

A18820-22 A18820-14 A18820-22 A18820-14
AAA18820-14EA 74.15 USD
AAA18820-14 CAAAA18820-22 AAA18820-22 CAAAA18820-14
Saponin
Saponin

A glycoside used to permeabilize cellular membranes

Melting Pt: 158 °C (dec.)
Density: 1,015…1.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00081981
CAS Number: 8047-15-2
EINECS: 232-462-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR