Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,5-Difluorophenol 97%

Supplier: Alfa Aesar
Danger

A18744-06 A18744-14 A18744-06 A18744-14
AAA18744-06EA 79.73 USD InStock
CAAAA18744-06 CAAAA18744-14 AAA18744-06 AAA18744-14
2,5-Difluorophenol 97%
2,5-Difluorophenol


Formula: C₆H₄F₂O
MW: 130.09 g/mol
Boiling Pt: 166 °C (1013 hPa)
Melting Pt: 42 °C
Flash Pt: 53 °C (127 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042501
CAS Number: 2713-31-7
EINECS: 000-000-0
UN: 2921
ADR: 8,II
Order Now

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty