1,4-Dicyclohexylbenzene 99%

Warning

A18708-14 A18708-06 A18708-22 A18708-14 A18708-22 A18708-06
AAAA18708-22EA 263.66 USD
CAAAAA18708-22 CAAAA18708-06 CAAAAA18708-14 AAAA18708-22 AAAA18708-14 AAA18708-06
1,4-Dicyclohexylbenzene 99%
1,4-Dicyclohexylbenzene
Formula: C₁₈H₂₆
MW: 242.4 g/mol
Boiling Pt: 192…195 °C (13 mmHg)
Melting Pt: 103…105 °C
Density: 1.049 g/cm³ (0 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001452
CAS Number: 1087-02-1
EINECS: 214-121-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR