2,5-Dimethoxyphenyl isothiocyanate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,4-Dimethoxy-2-isothiocyanatobenzene

A18630-18 A18630-09 A18630-09 A18630-18
AAA18630-09EA 163.99 USD
CAAAAA18630-18 CAAAA18630-09 AAA18630-09 AAAA18630-18
2,5-Dimethoxyphenyl isothiocyanate 98%
2,5-Dimethoxyphenyl isothiocyanate
Formula: C₉H₉NO₂S
Boiling Pt: 174…175 °C/10 mm
Melting Pt: 35…37 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041059
CAS Number: 40532-06-7
EINECS: 000-000-0
UN: 2923
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR