2-Bromo-4-(trifluoromethyl)aniline 97%

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: 4-Amino-3-bromo-α,α,α-trifluorotoluene 2-Bromo-α,α,α-trifluoro-p-toluidine

A18528-03 A18528-06 A18528-03 A18528-06
AAA18528-03EA 45 USD
CAAAA18528-03 AAA18528-03 AAA18528-06 CAAAA18528-06
2-Bromo-4-(trifluoromethyl)aniline 97%
2-Bromo-4-(trifluoromethyl)aniline
Formula: C₇H₅BrF₃N
MW: 240.02 g/mol
Boiling Pt: 109…110 °C/10 mm
Melting Pt: 26…33 °C
Density: 1.700
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042150
CAS Number: 57946-63-1
EINECS: 261-035-7
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR