2-Fluoro-3-(trifluoromethyl)benzonitrile 98%

Warning

Synonyms: 3-Cyano-2-fluorobenzotrifluoride

A18517-03 A18517-06 A18517-03 A18517-06
AAA18517-03EA 59.25 USD
CAAAA18517-06 AAA18517-03 AAA18517-06 CAAAA18517-03
2-Fluoro-3-(trifluoromethyl)benzonitrile 98%
2-Fluoro-3-(trifluoromethyl)benzonitrile
Formula: C₈H₃F₄N
MW: 189.11 g/mol
Boiling Pt: 212 °C (1013 hPa)
Melting Pt: 28 °C
Density: 1.323
Flash Pt: 85 °C(185 °F)
Storage Temperature: Ambient
MDL Number: MFCD00061152
CAS Number: 146070-35-1
UN: 3276
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR