1-Bromo-2,3,4-trifluorobenzene 99%

Supplier: Thermo Scientific Chemicals
Warning

A18514-06 A18514-03 A18514-03 A18514-06
AAA18514-03EA 26.36 USD
AAA18514-03 CAAAA18514-06 AAA18514-06 CAAAA18514-03
1-Bromo-2,3,4-trifluorobenzene 99%
1-Bromo-2,3,4-trifluorobenzene
Formula: C₆H₂BrF₃
MW: 210.98 g/mol
Boiling Pt: 47…49 °C/60 mm
Density: 1,777
Flash Pt: 62 °C(143 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013248
CAS Number: 176317-02-5
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR