4-Bromo-3-(trifluoromethyl)aniline 98%

Danger

Synonyms: 5-Amino-2-bromo-α,α,α-trifluorotoluene 4-Bromo-α,α,α-trifluoro-m-toluidine

A18380-14 A18380-06 A18380-03 A18380-03 A18380-06 A18380-14
AAA18380-06EA 53.8 USD
AAA18380-06 CAAAA18380-06 CAAAA18380-03 CAAAA18380-14 AAA18380-14 AAA18380-03
4-Bromo-3-(trifluoromethyl)aniline 98%
4-Bromo-3-(trifluoromethyl)aniline
Formula: C₇H₅BrF₃N
MW: 240.02 g/mol
Boiling Pt: 81…84 °C/0.5 mm
Melting Pt: 46…49 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007827
CAS Number: 393-36-2
EINECS: 206-885-1
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR