n-Heneicosane 99%

Supplier: Thermo Scientific Chemicals

A18198-06 A18198-03 A18198-06 A18198-14 A18198-14 A18198-03
AAA18198-14EA 323.99 USD
AAA18198-14 AAA18198-03 AAA18198-06 CAAAA18198-06 CAAAA18198-03 CAAAA18198-14
n-Heneicosane 99%
n-Heneicosane
Formula: H₃C(CH₂)₁₉CH₃
MW: 296.58 g/mol
Boiling Pt: 100 °C/2 mm
Melting Pt: 39…43 °C
Density: 0.792
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009346
CAS Number: 629-94-7
EINECS: 211-118-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR