4-Chloro-2-nitroaniline 98%

Supplier: Thermo Scientific Chemicals
Danger

A18143-0I A18143-0B A18143-0B A18143-30 A18143-30 A18143-0I
AAA18143-0IEA 1705.48 USD
CAAAA18143-30 AAA18143-0I CAAAA18143-0I AAA18143-0B CAAAA18143-0B AAA18143-30
4-Chloro-2-nitroaniline 98%
4-Chloro-2-nitroaniline
Formula: C₆H₅ClN₂O₂
MW: 172.57 g/mol
Melting Pt: 114…118 °C
Density: 1.37
Flash Pt: 191 °C(375 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007836
CAS Number: 89-63-4
EINECS: 201-925-4
UN: 2237
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR