1-(2,6-Dichlorophenyl)-2-thiourea 99%

Danger

Synonyms: 2,6-Dichlorophenylthiourea N-(2,6-Dichlorophenyl)thiourea

A18117-06 A18117-14 A18117-06 A18117-14
AAA18117-06EA 64.97 USD
AAA18117-06 AAA18117-14 CAAAA18117-06 CAAAA18117-14
1-(2,6-Dichlorophenyl)-2-thiourea 99%
1-(2,6-Dichlorophenyl)-2-thiourea
Formula: (C₆H₃Cl₂)NC(S)NH₂
MW: 221.11 g/mol
Boiling Pt: 326 °C (1013 hPa)
Melting Pt: 155…159 °C
Storage Temperature: Ambient
MDL Number: MFCD00041156
CAS Number: 6590-91-6
EINECS: 000-000-0
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR