Allylpentafluorobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Allyl-2,3,4,5,6-pentafluorobenzene3-(Pentafluorophenyl)-1-propene

A17834-14 A17834-06 A17834-14 A17834-06
AAA17834-14EA 162.99 USD
AAA17834-14 CAAAA17834-06 CAAAA17834-14 AAA17834-06
Allylpentafluorobenzene 98%
Allylpentafluorobenzene
Formula: (C₆F₅)CH₂CHCH₂
MW: 208.13 g/mol
Boiling Pt: 148…149 °C
Density: 1.358
Flash Pt: 42 °C(108 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000301
CAS Number: 1736-60-3
EINECS: 217-083-6
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR