2-Bromo-5-(trifluoromethyl)aniline 97%

Supplier: Thermo Scientific Chemicals

Danger

Synonyms: 3-Amino-4-bromo-α,α,α-trifluorotoluene 3-Amino-4-brom-α,α,α-trifluortoluol 6-Bromo-α,α,α-trifluoro-m-toluidine

A17810-03 A17810-03 A17810-06 A17810-14 A17810-14 A17810-06
AAA17810-03EA 25 USD
AAA17810-03 CAAAA17810-06 AAA17810-14 AAA17810-06 CAAAA17810-03 CAAAA17810-14
2-Bromo-5-(trifluoromethyl)aniline 97%
2-Bromo-5-(trifluoromethyl)aniline
Formula: C₇H₅BrF₃N
MW: 240.02 g/mol
Boiling Pt: 81…82 °C/5 mm
Melting Pt: 47…49 °C
Density: 1.675
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007637
CAS Number: 454-79-5
EINECS: 207-227-6
UN: 3267
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR