2-Chloro-3,5-dinitro-α,α,α-trifluorotoluene 98+%

Danger

Synonyms: 1-Chloro-2,4-dinitro-6-(trifluoromethyl)benzene 2-Chloro-3,5-dinitrobenzotrifluoride 2-Chloro-1,5-dinitro-3-(trifluoromethyl)benzene

A17617-06 A17617-06 A17617-14 A17617-14
AAA17617-14EA 126.69 USD
AAA17617-14 CAAAA17617-14 AAA17617-06 CAAAA17617-06
2-Chloro-3,5-dinitro-α,α,α-trifluorotoluene 98+%
2-Chloro-3,5-dinitro-α,α,α-trifluorotoluene
Formula: C₇H₂ClF₃N₂O₄
MW: 270.55 g/mol
Melting Pt: 61…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00007076
CAS Number: 392-95-0
EINECS: 206-883-0
UN: 2923
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR