n-Dotriacontane 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Dotriacontane

A17480-14 A17480-06 A17480-06 A17480-14
AAA17480-06EA 49.29 USD
AAA17480-06 CAAAA17480-06 AAA17480-14 CAAAA17480-14
n-Dotriacontane 98%
n-Dotriacontane
Formula: H₃C(CH₂)₃₀CH₃
MW: 450.88 g/mol
Boiling Pt: 300…310 °C
Melting Pt: 68…71 °C
Density: 0.812
Storage Temperature: Ambient
MDL Number: MFCD00009411
CAS Number: 544-85-4
EINECS: 208-881-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR