Hexachloro-1,3-butadiene 97%

Danger

A17316-30 A17316-0B A17316-18 A17316-18 A17316-30 A17316-0B
AAA17316-30EA 247.51 USD
CAAAA17316-0B AAA17316-30 AAA17316-0B CAAAA17316-18 AAA17316-18 CAAAA17316-30
Hexachloro-1,3-butadiene 97%
Hexachloro-1,3-butadiene
Formula: C₄Cl₆
MW: 260.76 g/mol
Boiling Pt: 215 °C
Melting Pt: –21 °C
Density: 1.680
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00000836
CAS Number: 87-68-3
EINECS: 201-765-5
UN: 2279
ADR: "6.1",III
Merck Index: 13,04697

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR