3-Chloro-2,4,5,6-tetrafluoropyridine 98%

Warning

Synonyms: 3-Chlorotetrafluoropyridine

A17285-14 A17285-14 A17285-06 A17285-06
AAA17285-06EA 44.75 USD
CAAAA17285-06 CAAAA17285-14 AAA17285-06 AAA17285-14
3-Chloro-2,4,5,6-tetrafluoropyridine 98%
3-Chloro-2,4,5,6-tetrafluoropyridine
Formula: C₅ClF₄N
MW: 185.51 g/mol
Boiling Pt: 118…121 °C
Density: 1.641
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00010624
CAS Number: 1735-84-8
UN: 2810
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR