1,2-Diethoxybenzene 98%

A17283-30 A17283-18 A17283-18 A17283-30
AAA17283-30EA 267 USD
AAA17283-30 AAA17283-18 CAAAA17283-30 CAAAA17283-18
1,2-Diethoxybenzene 98%
1,2-Diethoxybenzene
Formula: C₁₀H₁₄O₂
MW: 166.22 g/mol
Boiling Pt: 218…220 °C
Melting Pt: 43…45 °C
Storage Temperature: Ambient
MDL Number: MFCD00015143
CAS Number: 2050-46-6
EINECS: 218-089-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR