3,4-Dichloroanisole 98%

Warning

A17242-14 A17242-06 A17242-06 A17242-14
AAA17242-06EA 59.81 USD
AAA17242-06 AAA17242-14 CAAAA17242-14 CAAAA17242-06
3,4-Dichloroanisole 98%
3,4-Dichloroanisole
Formula: C₇H₆Cl₂O
MW: 177.03 g/mol
Boiling Pt: 200…204 °C
Melting Pt: –8 °C
Density: 1.330
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00070785
CAS Number: 36404-30-5
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR