Dibromochloromethane 98%, rods

Supplier: Thermo Scientific Chemicals
Warning

A16938-06 A16938-22 A16938-14 A16938-14 A16938-22 A16938-06
AAA16938-06EA 34.09 USD
CAAAA16938-22 AAA16938-06 AAA16938-14 CAAAA16938-06 AAA16938-22 CAAAA16938-14
Dibromochloromethane 98%, rods
Dibromochloromethane
Formula: Br₂ClCH
MW: 208.28 g/mol
Boiling Pt: 115…118 °C
Melting Pt: –22 °C
Density: 2.420
Storage Temperature: Refrigerator
MDL Number: MFCD00000820
CAS Number: 124-48-1
EINECS: 204-704-0
Merck Index: 13,02154

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR