2'-Fluoroacetanilide 98+%

Warning

Synonyms: N-(2-Fluorophenyl)acetamide

A16927-14 A16927-14 A16927-22 A16927-22
AAA16927-14EA 57.39 USD
CAAAA16927-22 AAA16927-14 AAA16927-22 CAAAA16927-14
2'-Fluoroacetanilide 98+%
2'-Fluoroacetanilide
Formula: C₈H₈FNO
MW: 153.16 g/mol
Boiling Pt: 140…142 °C/14 mm
Melting Pt: 76…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00000315
CAS Number: 399-31-5
EINECS: 206-916-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR