DL-Valine 99%

Supplier: Thermo Scientific Chemicals

Synonyms: (±)-Valine H-DL-Val-OH

A16756-22 A16756-22 A16756-36 A16756-0E A16756-36 A16756-0E
AAA16756-0EEA 553.99 USD
AAA16756-0E CAAAA16756-36 CAAAA16756-0E CAAAA16756-22 AAA16756-22 AAA16756-36
DL-Valine 99%
DL-Valine
Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Boiling Pt: 217 °C (1013 hPa)
Melting Pt: ca 296 °C (subl.)
Density: 1.352 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004267
CAS Number: 516-06-3
EINECS: 208-220-0
Merck Index: 12,10046

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR