Ethyl-2-fluorobenzoate 98+%

Synonyms: 2-Fluorobenzoic acid ethyl ester

A16564-22 A16564-14 A16564-22 A16564-14
AAA16564-14EA 63.89 USD
AAA16564-14 AAA16564-22 CAAAA16564-22 CAAAA16564-14
Ethyl-2-fluorobenzoate 98+%
Ethyl 2-fluorobenzoate
Formula: C₉H₉FO₂
MW: 168.17 g/mol
Boiling Pt: 217…220 °C
Storage Temperature: Ambient
MDL Number: MFCD00039214
CAS Number: 443-26-5
EINECS: 207-134-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR