1-Bromo-3-(trifluoromethoxy)benzene 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(Trifluoromethoxy)bromobenzene

A16561-06 A16561-06 A16561-03 A16561-03 A16561-14 A16561-14
AAA16561-14EA 324.99 USD
AAA16561-14 AAAA16561-03 AAA16561-06 CAAAA16561-06 CAAAAA16561-03 CAAAA16561-14
1-Bromo-3-(trifluoromethoxy)benzene 98+%
1-Bromo-3-(trifluoromethoxy)benzene
Formula: C₇H₄BrF₃O
MW: 241.01 g/mol
Boiling Pt: 156…158 °C
Density: 1,620
Flash Pt: 62 °C(143 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040943
CAS Number: 2252-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR