1-(3,5-Dichlorophenyl)-2-thiourea 98+%

Danger

Synonyms: 3,5-Dichlorophenylthiourea N-(3,5-Dichlorophenyl)thiourea

A16558-06 A16558-14 A16558-03 A16558-06 A16558-03 A16558-14
AAA16558-06EA 101.56 USD
AAA16558-06 AAA16558-03 AAA16558-14 CAAAA16558-14 CAAAA16558-03 CAAAA16558-06
1-(3,5-Dichlorophenyl)-2-thiourea 98+%
1-(3,5-Dichlorophenyl)-2-thiourea
Formula: (C₆H₃Cl₂)NC(S)NH₂
MW: 221.11 g/mol
Boiling Pt: 326 °C (1013 hPa)
Melting Pt: 175…179 °C
Storage Temperature: Ambient
MDL Number: MFCD00041168
CAS Number: 107707-33-5
EINECS: 000-000-0
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR