Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 4pm ET Saturday through 8pm Sunday.

1,2,3-Trifluoro-4-nitrobenzene 97%

Supplier: Alfa Aesar
Danger
Synonyms: 2,3,4-Trifluoronitrobenzene

A16313-14 A16313-22 A16313-06 A16313-22 A16313-14 A16313-06
AAA16313-14EA 107.12 USD InStock
AAA16313-14 AAA16313-22 CAAAA16313-06 CAAAA16313-22 CAAAA16313-14 AAA16313-06
1,2,3-Trifluoro-4-nitrobenzene 97%
1,2,3-Trifluoro-4-nitrobenzene


Formula: C₆H₂F₃NO₂
MW: 177.08 g/mol
Boiling Pt: 91…93 °C (20 mmHg)
Density: 1.53 g/cm³ (20 °C)
Flash Pt: 93 °C (199 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041546
CAS Number: 771-69-7
EINECS: 212-238-4
UN: 2810
ADR: 6.1,III
Order Now

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty