1-Ethylpiperidine 99%

Supplier: Thermo Scientific Chemicals
Danger

A16294-36 A16294-36
AAA16294-36EA 116.63 USD
AAA16294-36 CAAAA16294-36
1-Ethylpiperidine 99%
1-Ethylpiperidine
Formula: C₇H₁₅N
MW: 113.2 g/mol
Boiling Pt: 129…131 °C
Melting Pt: –20 °C
Density: 0.825
Flash Pt: 18 °C(64 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006507
CAS Number: 766-09-6
EINECS: 212-161-6
UN: 2386
ADR: "3",II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR