4-Bromo-2-fluorocinnamic acid 98%

Warning

A16028-14 A16028-06 A16028-06 A16028-14
AAA16028-14EA 159.5 USD
AAA16028-14 AAA16028-06 CAAAA16028-14 CAAAA16028-06
4-Bromo-2-fluorocinnamic acid 98%
4-Bromo-2-fluorocinnamic acid
Formula: C₉H₆BrFO₂
MW: 245.05 g/mol
Boiling Pt: 322 °C (1013 hPa)
Melting Pt: 216…221 °C
Storage Temperature: Ambient
MDL Number: MFCD00143267
CAS Number: 149947-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR