N-Ethylphthalimide 98%

Supplier: Thermo Scientific Chemicals
Warning

A16014-14 A16014-06 A16014-14 A16014-22 A16014-06 A16014-22
AAAA16014-22EA 1078.96 USD
AAAA16014-22 AAA16014-06 CAAAA16014-06 AAA16014-14 CAAAAA16014-22 CAAAA16014-14
N-Ethylphthalimide 98%
N-Ethylphthalimide
Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Boiling Pt: 286 °C (1013 hPa)
Melting Pt: 74…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00014583
CAS Number: 5022-29-7
EINECS: 225-707-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR