4-(Benzyloxy)benzaldehyde 98%

A15934-22 A15934-36 A15934-22 A15934-14 A15934-14 A15934-36
AAA15934-36EA 421.27 USD
CAAAA15934-14 CAAAA15934-36 CAAAA15934-22 AAA15934-36 AAA15934-14 AAA15934-22
4-(Benzyloxy)benzaldehyde 98%
4-(Benzyloxy)benzaldehyde
Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
Boiling Pt: 197…199 °C/20 mm
Melting Pt: 69…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00003387
CAS Number: 4397-53-9
EINECS: 224-527-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR