2-(Trifluoromethoxy)benzonitrile 97%

Warning

A15710-06 A15710-03 A15710-06 A15710-03
AAA15710-03EA 53.94 USD
CAAAA15710-06 AAA15710-03 CAAAA15710-03 AAA15710-06
2-(Trifluoromethoxy)benzonitrile 97%
2-(Trifluoromethoxy)benzonitrile
Formula: C₈H₄F₃NO
MW: 187.12 g/mol
Boiling Pt: 80…82 °C/15 mm
Density: 1.245
Storage Temperature: Ambient
MDL Number: MFCD00042409
CAS Number: 63968-85-4
EINECS: 264-584-0
UN: 3276
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR