2-Methoxyfuran 97%

Supplier: Thermo Scientific Chemicals
Danger

A15593-03 A15593-06 A15593-14 A15593-14 A15593-03 A15593-06
AAA15593-03EA 97.29 USD
AAA15593-03 AAA15593-14 CAAAA15593-14 CAAAA15593-03 CAAAA15593-06 AAA15593-06
2-Methoxyfuran 97%
2-Methoxyfuran
Formula: C₅H₆O₂
Boiling Pt: 110…111 °C
Density: 1.065
Flash Pt: 10 °C(50 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00003227
CAS Number: 25414-22-6
EINECS: 246-953-8
UN: 1992
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR