N-Methyl-4-nitroaniline 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-(Methylamino)-4-nitrobenzene
A15548-06
A15548-22
A15548-14
AAA15548-22EA
461.99
USD
AAA15548-22
AAA15548-06
AAA15548-14
N-Methyl-4-nitroaniline 97%
N-Methyl-4-nitroaniline
Formula:
C₇H₈N₂O₂ Boiling Pt: 291 °C (1013 hPa) Melting Pt: 147…154 °C Density: 1.201 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007305 CAS Number: 100-15-2 EINECS: 202-823-2 UN: 2811 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...