3-Methyl-4-nitrobenzoic acid 99%

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: 3-Methyl-4-nitrobenzoic acid

A15523-22 A15523-22 A15523-36 A15523-36
AAAA15523-36EA 176 USD
AAAA15523-36 CAAAAA15523-36 CAAAA15523-22 AAA15523-22
3-Methyl-4-nitrobenzoic acid 99%
4-Nitro-m-toluic acid
Formula: C₈H₇NO₄
MW: 181.15 g/mol
Boiling Pt: 356 °C (1013 hPa)
Melting Pt: 215…220 °C
Storage Temperature: Ambient
MDL Number: MFCD00007168
CAS Number: 3113-71-1
EINECS: 221-479-4
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR