2-Ethylfuran 98+%

Supplier: Thermo Scientific Chemicals
Danger

A15520-22 A15520-06 A15520-14 A15520-06 A15520-14 A15520-22
AAA15520-14EA 187.99 USD
AAA15520-14 AAA15520-06 CAAAA15520-22 CAAAA15520-14 AAA15520-22 CAAAA15520-06
2-Ethylfuran 98+%
2-Ethylfuran
Formula: C₆H₈O
Boiling Pt: 92…93 °C
Density: 0.912
Flash Pt: -2 °C(28 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003259
CAS Number: 3208-16-0
EINECS: 221-714-0
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR