2,3-Dichlorothiophenol 97%

Warning

Synonyms: 1,2-Dichloro-3-mercaptobenzene 2,3-Dichlorobenzene-1-thiol 2,3-Dichlorobenzenethiol

A15491-22 A15491-22 A15491-14 A15491-06 A15491-14 A15491-06
AAA15491-14EA 206.69 USD
AAA15491-14 AAAA15491-22 CAAAA15491-06 CAAAA15491-14 AAA15491-06 CAAAAA15491-22
2,3-Dichlorothiophenol 97%
2,3-Dichlorothiophenol
Formula: C₆H₄Cl₂S
MW: 179.07 g/mol
Boiling Pt: 95…98 °C/0.5 mm
Melting Pt: 55…58 °C
Storage Temperature: Ambient
MDL Number: MFCD00041420
CAS Number: 17231-95-7
EINECS: 000-000-0
UN: 3335
ADR: "9",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR