1,4-Diethoxybenzene 98%

Supplier: Thermo Scientific Chemicals

A15385-36 A15385-14 A15385-14 A15385-22 A15385-36 A15385-22
AAA15385-22EA 122.99 USD
CAAAA15385-22 CAAAA15385-36 CAAAA15385-14 AAA15385-22 AAA15385-14 AAA15385-36
1,4-Diethoxybenzene 98%
1,4-Diethoxybenzene
Formula: C₁₀H₁₄O₂
MW: 166.22 g/mol
Boiling Pt: 245…248 °C
Melting Pt: 69…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00015146
CAS Number: 122-95-2
EINECS: 204-585-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR