3,5-Diiodoanthranilic acid 98+%

Warning

Synonyms: 2-Amino-3,5-diiodobenzoic acid

A15308-06 A15308-14 A15308-22
CAAAA15308-06 CAAAA15308-14 CAAAA15308-22
3,5-Diiodoanthranilic acid 98+%
3,5-Diiodoanthranilic acid
Formula: C₇H₅I₂NO₂
MW: 388.93 g/mol
Boiling Pt: 410 °C (1013 hPa)
Melting Pt: ca 240 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007682
CAS Number: 609-86-9
EINECS: 210-201-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR