2,3,4,5,6-Pentamethylbenzophenone 98%

A15182-06 A15182-14 A15182-06 A15182-14
AAA15182-06EA 72.67 USD
CAAAA15182-06 CAAAA15182-14 AAA15182-06 AAA15182-14
2,3,4,5,6-Pentamethylbenzophenone 98%
2,3,4,5,6-Pentamethylbenzophenone
Formula: C₁₈H₂₀O
MW: 252.36 g/mol
Melting Pt: 137…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00051595
CAS Number: 20386-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR