N-Ethylaniline 97%
|
Danger
|
Synonyms:
N-Ethyl-N-phenylamine
A15060-AP
A15060-0F
A15060-AP
A15060-0F
AAA15060-APEA
33.17
USD
CAAAA15060-AP
AAA15060-AP
CAAAA15060-0F
AAA15060-0F
N-Ethylaniline 97%
N-Ethylaniline
Formula:
C₈H₁₁N MW: 121.18 g/mol Boiling Pt: 204…205 °C Melting Pt: –64 °C Density: 0.961 Flash Pt: 85 °C(185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009025 CAS Number: 103-69-5 EINECS: 203-135-5 UN: 2272 ADR: "6.1",III Merck Index: 13,03800 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...