N-(4-Chlorophenyl)phthalimide 98+%

Warning

Synonyms: 2-(4-Chlorophenyl)-1H-isoindole-1,3(2H)-dione

A15057-22 A15057-14 A15057-22 A15057-06 A15057-14 A15057-06
AAA15057-22EA 325.23 USD
CAAAA15057-06 AAA15057-22 AAA15057-14 CAAAA15057-14 CAAAA15057-22 AAA15057-06
N-(4-Chlorophenyl)phthalimide 98+%
N-(4-Chlorophenyl)phthalimide
Formula: C₁₄H₈ClNO₂
MW: 257.68 g/mol
Melting Pt: 195…199 °C
Storage Temperature: Ambient
MDL Number: MFCD00023048
CAS Number: 7386-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR