5-Iodoisatin 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 5-Iodo-1H-indole-2,3-dione 5-Iodoindoline-2,3-dione

A14988-06 A14988-14 A14988-06 A14988-14
AAA14988-14EA 309.07 USD
CAAAA14988-14 AAA14988-14 AAA14988-06 CAAAA14988-06
5-Iodoisatin 97%
5-Iodoisatin
Formula: C₈H₄INO₂
MW: 273.03 g/mol
Melting Pt: ca 276 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00016899
CAS Number: 20780-76-1
EINECS: 244-035-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR