6-Chlorochroman-4-one 99%

A14975-06 A14975-03 A14975-03 A14975-06
AAA14975-03EA 50.89 USD
CAAAA14975-03 AAA14975-03 AAA14975-06 CAAAA14975-06
6-Chlorochroman-4-one 99%
6-Chlorochroman-4-one
Formula: C₉H₇ClO₂
MW: 182.61 g/mol
Melting Pt: 101…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00024070
CAS Number: 37674-72-9
EINECS: 253-611-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR