1-Bromoadamantane 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
1-Adamantyl bromide
A14893-14
A14893-22
A14893-14
A14893-22
AAA14893-22EA
133
USD
AAA14893-22
CAAAA14893-14
CAAAA14893-22
AAA14893-14
1-Bromoadamantane 99%
1-Bromoadamantane
Formula:
C₁₀H₁₅Br MW: 215.13 g/mol Melting Pt: 117…121 °C Storage Temperature: Ambient |
MDL Number:
MFCD00074721 CAS Number: 768-90-1 EINECS: 212-200-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...