Dichloroacetic acid (DCA) 99%

Danger

Synonyms: DCA

A14740-22 A14740-0E A14740-0E
AAAA14740-22EA 39.52 USD
AAAA14740-22 AAA14740-0E CAAAA14740-0E
Dichloroacetic acid (DCA) 99%
Dichloroacetic acid (DCA)
Formula: C₂H₂Cl₂O₂
MW: 128.94 g/mol
Boiling Pt: 192…194 °C
Melting Pt: 9…11 °C
Density: 1.564
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004223
CAS Number: 79-43-6
EINECS: 201-207-0
UN: 1764
ADR: "8",II
Merck Index: 13,03075

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR