Pentamethylbenzonitrile 98%

Warning

A14733-06 A14733-03 A14733-03 A14733-06
AAA14733-06EA 111.07 USD
CAAAA14733-03 AAA14733-06 CAAAA14733-06 AAA14733-03
Pentamethylbenzonitrile 98%
Pentamethylbenzonitrile
Formula: C₁₂H₁₅N
MW: 173.26 g/mol
Boiling Pt: 290…292 °C (1013 hPa)
Melting Pt: 158…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00015450
CAS Number: 5144-10-5
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR