2-Fluoro-6-phenoxybenzonitrile 95%

Warning

A14566-06 A14566-06 A14566-03 A14566-03
AAA14566-06EA 126.66 USD
AAA14566-06 AAA14566-03 CAAAA14566-06 CAAAA14566-03
2-Fluoro-6-phenoxybenzonitrile 95%
2-Fluoro-6-phenoxybenzonitrile
Formula: C₁₃H₈FNO
MW: 213.21 g/mol
Boiling Pt: 338 °C (1013 hPa)
Melting Pt: 90…93 °C
Storage Temperature: Ambient
MDL Number: MFCD00068201
CAS Number: 175204-06-5
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR