4-Propylbenzoic acid 98+%

Warning

A14538-09 A14538-04 A14538-18 A14538-09 A14538-04 A14538-18
AAA14538-18EA 96.29 USD
AAA14538-18 CAAAA14538-18 AAA14538-04 CAAAA14538-09 CAAAA14538-04 AAA14538-09
4-Propylbenzoic acid 98+%
4-Propylbenzoic acid
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 298 °C (1013 hPa)
Melting Pt: 139…144 °C
Density: ∼1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013996
CAS Number: 2438-05-3
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR